Novus Biologicals
Manufacturer Code:NBP246283
Catalog # NBP246283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5C7 |
Immunogen | Full length human recombinant protein of human CHMP5 (NP_057494) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: apoptosis-related protein PNAS-2; apoptosis-related protein PNAS-2 C9orf83 charged multivesicular body protein 5 chromatin modifying protein 5 Chromatin-modifying protein 5 chromosome 9 open reading frame 83 HSPC177 hVps60 PNAS-2 SNF7 domain containing 2 SNF7 domain-containing protein 2 SNF7DC2 Vacuolar protein sorting-associated protein 60 Vps60CGI-34; Charged multivesicular body protein 5; chromatin modifying protein 5; Chromatin-modifying protein 5; hVps60; SNF7 domain containing 2; SNF7 domain-containing protein 2; Vacuolar protein sorting-associated protein 60; Vps60
Gene Aliases: C9orf83; CGI-34; CHMP5; HSPC177; PNAS-114; PNAS-2; SNF7DC2; Vps60
UniProt ID: (Human) Q9NZZ3
Entrez Gene ID: (Human) 51510
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.