Invitrogen
Manufacturer Code:PA532050
Catalog # PIPA532050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 168 of Human CHMP4C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Charged multivesicular body protein 4c; CHMP4c; chromatin modifying protein 4C; Chromatin-modifying protein 4c; hSnf7-3; hVps32-3; SNF7 homolog associated with Alix 3; Snf7 homologue associated with Alix 3; SNF7-3; SNF7-3 Shax3 VPS32C SNF7 homolog associated with Alix 3 Snf7 homologue associated with Alix 3 charged multivesicular body protein 4c chromatin modifying protein 4C chromatin-modifying protein 4c hSnf7-3 hVps32-3 vacuolar protein sorting-associated protein 32-3 vps32-3; Vacuolar protein sorting-associated protein 32-3; Vps32-3
Gene Aliases: 2010012P02Rik; 2210015K02Rik; 2310010I16Rik; CHMP4C; SHAX3; SNF7-3; VPS32C
UniProt ID: (Human) Q96CF2, (Mouse) Q9D7F7
Entrez Gene ID: (Human) 92421, (Mouse) 66371
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.