Invitrogen
Manufacturer Code:PA5114255
Catalog # PIPA5114255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal region of Mouse Chmp4c |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010012P02Rik 2210015K02Rik 2310010I16Rik Charged multivesicular body protein 4c CHMP4C chromatin modifying protein 4C Chromatin-modifying protein 4c hSnf7-3 hVps32-3 SHAX3 SNF7 homolog associated with Alix 3 Snf7 homologue associated with Alix 3 SNF7-3 Vacuolar protein sorting-associated protein 32-3 Vps32-3 VPS32C; Charged multivesicular body protein 4c; CHMP4c; chromatin modifying protein 4C; Chromatin-modifying protein 4c; Snf7 homologue associated with Alix 3
Gene Aliases: 2010012P02Rik; 2210015K02Rik; 2310010I16Rik; Chmp4c; Shax3; Snf7-3
UniProt ID: (Mouse) Q9D7F7
Entrez Gene ID: (Mouse) 66371
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.