Novus Biologicals
Manufacturer Code:NBP215888
Catalog # NBP215888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CHMP4C. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Charged multivesicular body protein 4c; charged multivesicular body protein 4c CHMP4c chromatin modifying protein 4C Chromatin-modifying protein 4c hSnf7-3 hVps32-3 MGC22825 Shax3 SNF7 homolog associated with Alix 3 Snf7 homologue associated with Alix 3 SNF7-3 Vacuolar protein sorting-associated protein 32-3 Vps32-3 VPS32C; CHMP4c; chromatin modifying protein 4C; Chromatin-modifying protein 4c; hSnf7-3; hVps32-3; SNF7 homolog associated with Alix 3; Snf7 homologue associated with Alix 3; SNF7-3; Vacuolar protein sorting-associated protein 32-3; Vps32-3
Gene Aliases: CHMP4C; SHAX3; SNF7-3; VPS32C
UniProt ID: (Human) Q96CF2
Entrez Gene ID: (Human) 92421
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.