OriGene
Manufacturer Code:TA890004
Catalog # 50-168-4720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from C terminal of Human CHMP4C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Charged multivesicular body protein 4c; CHMP4c; chromatin modifying protein 4C; Chromatin-modifying protein 4c; hSnf7-3; hVps32-3; SNF7 homolog associated with Alix 3; Snf7 homologue associated with Alix 3; SNF7-3; SNF7-3 Shax3 VPS32C; Vacuolar protein sorting-associated protein 32-3; Vps32-3
Gene Aliases: CHMP4C; SHAX3; SNF7-3; VPS32C
UniProt ID: (Human) Q96CF2
Entrez Gene ID: (Human) 92421
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.