Abnova
Manufacturer Code:H00092421K
Catalog # 89-208-998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CHMP4C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CHMP4C peptide using ARM Technology.
For Research Use Only
Protein Aliases: Charged multivesicular body protein 4c; CHMP4c; chromatin modifying protein 4C; Chromatin-modifying protein 4c; hSnf7-3; hVps32-3; MGC22825 SNF7-3 Shax3; SNF7 homolog associated with Alix 3; Snf7 homologue associated with Alix 3; SNF7-3; Vacuolar protein sorting-associated protein 32-3; Vps32-3
Gene Aliases: CHMP4C; SHAX3; SNF7-3; VPS32C
UniProt ID: (Human) Q96CF2
Entrez Gene ID: (Human) 92421
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.