Novus Biologicals
Manufacturer Code:NBP287183
Catalog # NB034805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human CHMP4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf178 charged multivesicular body protein 4b CHMP4A CHMP4b chromatin modifying protein 4B Chromatin-modifying protein 4b chromosome 20 open reading frame 178 dJ553F4.4 hSnf7-2 hVps32-2 SHAX1 SNF7 SNF7 homolog associated with Alix 1 Snf7 homologue associated with Alix 1 SNF7-2CTPP3 Vacuolar protein sorting-associated protein 32-2 vacuolar protein-sorting-associated protein 32-2 Vps32-2 VPS32B; Charged multivesicular body protein 4b; CHMP4b; chromatin modifying protein 4B; Chromatin-modifying protein 4b; hSnf7-2; hVps32-2; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; SNF7-2; Vacuolar protein sorting-associated protein 32-2; vacuolar protein-sorting-associated protein 32-2; Vps32-2
Gene Aliases: C20orf178; CHMP4A; CHMP4B; CTPP3; CTRCT31; dJ553F4.4; SHAX1; SNF7; SNF7-2; Vps32-2; VPS32B
UniProt ID: (Human) Q9H444
Entrez Gene ID: (Human) 128866
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.