Proteintech
Manufacturer Code:136831AP150UL
Catalog # 50-172-8932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHMP4B Fusion Protein Ag4544 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf178 CHMP4B chromatin modifying protein 4B CTPP3 hSnf7 2 hVps32 2 Shax1 SNF7 SNF7 2 Vps32 2; Charged multivesicular body protein 4b; CHMP4b; chromatin modifying protein 4B; Chromatin-modifying protein 4b; hSnf7-2; hVps32-2; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; SNF7-2; Vacuolar protein sorting-associated protein 32-2; vacuolar protein-sorting-associated protein 32-2; Vps32-2
Gene Aliases: 2010012F05Rik; C20orf178; C76846; CHMP4A; CHMP4B; CTPP3; CTRCT31; dJ553F4.4; RGD1309846; RGD1565889; SHAX1; SNF7; SNF7-2; Vps32-2; VPS32B
UniProt ID: (Human) Q9H444, (Mouse) Q9D8B3
Entrez Gene ID: (Human) 128866, (Mouse) 75608, (Rat) 100359642
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.