Invitrogen
Manufacturer Code:PA570553
Catalog # PIPA570553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human CHMP2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Charged multivesicular body protein 2a; CHMP2a; chromatin modifying protein 2A; Chromatin-modifying protein 2a; mVps2; putative breast adenocarcinoma marker; putative breast adenocarcinoma marker (32kD); Putative breast adenocarcinoma marker BC-2; Vacuolar protein sorting-associated protein 2; Vacuolar protein sorting-associated protein 2-1; vacuolar protein-sorting-associated protein 2; VPS2 homolog A; Vps2-1
Gene Aliases: 1500016L11Rik; BC-2; BC2; CHMP2; CHMP2A; fj10d05; RGD1305050; VPS2; VPS2A; wu:fi18h03; wu:fj10d05; zgc:55361
UniProt ID: (Human) O43633, (Mouse) Q9DB34, (Zebrafish) Q7ZW25
Entrez Gene ID: (Dog) 476364, (Bovine) 523847, (Human) 27243, (Horse) 100064860, (Rabbit) 100340798, (Rat) 365191, (Mouse) 68953, (Guinea pig) 100717259, (Zebrafish) 798901
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.