Invitrogen
Manufacturer Code:PA559831
Catalog # PIPA559831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CHMP2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Charged multivesicular body protein 2a; CHMP2a; chromatin modifying protein 2A; Chromatin-modifying protein 2a; putative breast adenocarcinoma marker (32kD); Putative breast adenocarcinoma marker BC-2; Vacuolar protein sorting-associated protein 2-1; VPS2 homolog A; Vps2-1
Gene Aliases: BC-2; BC2; CHMP2; CHMP2A; VPS2; VPS2A
UniProt ID: (Human) O43633
Entrez Gene ID: (Human) 27243
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.