Invitrogen
Manufacturer Code:PA5106311
Catalog # PIPA5106311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CHMP2A corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500016L11Rik BC2 BC-2 Charged multivesicular body protein 2a CHMP2 CHMP2a chromatin modifying protein 2A Chromatin-modifying protein 2a mVps2 putative breast adenocarcinoma marker (32kD) Putative breast adenocarcinoma marker BC-2 RGD1305050 Vacuolar protein sorting-associated protein 2 vacuolar protein sorting-associated protein 2-1 vacuolar protein-sorting-associated protein 2 VPS2 VPS2 homolog A Vps2-1 VPS2A; Charged multivesicular body protein 2a; CHMP2a; chromatin modifying protein 2A; Chromatin-modifying protein 2a; mVps2; putative breast adenocarcinoma marker (32kD); Putative breast adenocarcinoma marker BC-2; Vacuolar protein sorting-associated protein 2; Vacuolar protein sorting-associated protein 2-1; vacuolar protein-sorting-associated protein 2; VPS2 homolog A; Vps2-1
Gene Aliases: 1500016L11Rik; BC-2; BC2; CHMP2; CHMP2A; RGD1305050; VPS2; VPS2A
UniProt ID: (Human) O43633, (Mouse) Q9DB34
Entrez Gene ID: (Human) 27243, (Rat) 365191, (Mouse) 68953
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.