Invitrogen
Manufacturer Code:PA5100436
Catalog # PIPA5100436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CHMP1A corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900018H07Rik charged multivesicular body protein 1/chromatin modifying protein 1 charged multivesicular body protein 1a Chmp1 Chmp1a chromatin modifying protein 1A chromatin-modifying protein 1a hVps46-1 KIAA0047 PCH8 PCOLN3 procollagen (type III) N-endopeptidase protease metallo 1 33kD PRSM1 Vacuolar protein sorting-associated protein 46-1 Vps46-1 VPS46A; charged multivesicular body protein 1/chromatin modifying protein 1; Charged multivesicular body protein 1a; CHMP1a; chromatin modifying protein 1A; Chromatin-modifying protein 1a; procollagen (type III) N-endopeptidase; protease, metallo, 1, 33kD; Vacuolar protein sorting-associated protein 46-1; Vps46-1
Gene Aliases: 2900018H07Rik; CHMP1; CHMP1A; KIAA0047; PCH8; PCOLN3; PRSM1; VPS46-1; VPS46A
UniProt ID: (Human) Q9HD42, (Mouse) Q921W0
Entrez Gene ID: (Human) 5119, (Mouse) 234852
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.