Invitrogen
Manufacturer Code:PA536289
Catalog # PIPA536289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 300-342 of human ChoKB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Choline kinase beta; Choline kinase-like protein; Choline/ethanolamine kinase; choline/ethanolamine kinase beta; Choline/ethanolamine kinase Choline kinase beta Short name&equalsCK CKB Choline kinase-like protein Ethanolamine kinase Short name&equalsEK Ethanolamine kinase beta EKB choline/ethanolamine kinase beta CKEKB CHKB CHETK CHKL; choline/ethanolamine kinase-b; choline/ethanolaminekinase; CK; CKB; CKEKB; EK; EKB; Ethanolamine kinase; Ethanolamine kinase beta
Gene Aliases: CHETK; CHKB; CHKL; CK; Ck/Ek; Ck/Ek-beta; CKB; CKEKB; EK; EKB; MDCMC
UniProt ID: (Human) Q9Y259, (Rat) O54783, (Mouse) O55229
Entrez Gene ID: (Human) 1120, (Rat) 29367, (Mouse) 12651
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.