Invitrogen
Manufacturer Code:PA5112385
Catalog # PIPA5112385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Choline/ethanolamine kinase protein (2-99AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chetk Chkb Chkl Choline kinase beta Choline kinase-like protein Choline/ethanolamine kinase choline/ethanolamine kinase beta choline/ethanolamine kinase-b choline/ethanolaminekinase CK Ck/Ek Ck/Ek-beta CKB CKEKB EK EKB Ethanolamine kinase Ethanolamine kinase beta MDCMC; Choline kinase beta; Choline kinase-like protein; Choline/ethanolamine kinase; choline/ethanolamine kinase beta; choline/ethanolamine kinase-b; choline/ethanolaminekinase; CK; CKB; CKEKB; EK; EKB; Ethanolamine kinase; Ethanolamine kinase beta
Gene Aliases: CHETK; CHKB; CHKL; CK; Ck/Ek; Ck/Ek-beta; CKB; CKEKB; EK; EKB; MDCMC
UniProt ID: (Human) Q9Y259, (Rat) O54783, (Mouse) O55229
Entrez Gene ID: (Human) 1120, (Rat) 29367, (Mouse) 12651
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.