Invitrogen
Manufacturer Code:PA515503
Catalog # PIPA515503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 427-457 amino acids from the C-terminal region of human Choline kinase alpha (CHK) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHETK-alpha; Choline kinase alpha; Choline kinase alpha CK CHETK-alpha CHKA CHK CKI; CK; EK; Ethanolamine kinase
Gene Aliases: CHK; CHKA; CK; CKI; EK
UniProt ID: (Human) P35790
Entrez Gene ID: (Human) 1119
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.