Invitrogen
Manufacturer Code:PA513816
Catalog # PIPA513816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 64-94 amino acids from the N-terminal region of human CHK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHETK-alpha; Choline kinase alpha; Choline kinase alpha CK CHETK-alpha; choline/ethanolamine kinase alpha; CK; EK; Ethanolamine kinase
Gene Aliases: Chetk-alpha; CHK; CHKA; ChoK; CK; CK/EK-alpha; CKI; EK; EtnK-alpha
UniProt ID: (Human) P35790, (Mouse) O54804
Entrez Gene ID: (Human) 1119, (Mouse) 12660
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.