Invitrogen
Manufacturer Code:PA5109529
Catalog # PIPA5109529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human CHKA (NP_001268.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHETK-alpha; CHETK-alpha CHK Chka ChoK Choline kinase alpha CK CK/EK-alpha CKI Ckr CK-R EK Ethanolamine kinase EtnK-alpha; Choline kinase alpha; choline kinase R; choline/ethanolamine kinase alpha; CK; EK; Ethanolamine kinase
Gene Aliases: Chetk-alpha; CHK; CHKA; ChoK; CK; CK-R; CK/EK-alpha; CKI; Ckr; EK; EtnK-alpha
UniProt ID: (Human) P35790, (Mouse) O54804, (Rat) Q01134
Entrez Gene ID: (Human) 1119, (Mouse) 12660, (Rat) 29194
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.