Invitrogen
Manufacturer Code:PA5109528
Catalog # PIPA5109528
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 217-466 of human CHIT1 (NP_003456.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2300002L19Rik CHI3 CHIT CHIT1 CHITD Chitinase 1 Chitinase-1 Chitotriosidase Chitotriosidase-1 EC 3.2.1.14 FLJ00314 MGC125322; chitinase 1 (chitotriosidase); Chitinase-1; Chitotriosidase-1; plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase
Gene Aliases: 2300002L19Rik; CHI3; CHIT; CHIT1; CHITD
UniProt ID: (Human) Q13231, (Mouse) Q9D7Q1
Entrez Gene ID: (Human) 1118, (Mouse) 71884
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.