Invitrogen
Manufacturer Code:PA547323
Catalog # PIPA547323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Chitinase 3-like 3/ECF-L Tyr22-Tyr398 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-N-acetylhexosaminidase Ym1; CHI3L3 ECF-L Ym1 AI505981 Chi3l3 ECF-L Ym1; chitinase 3-like 3; Chitinase-3-like protein 3; Chitinase-like protein 3; ECF-L; Eosinophil chemotactic cytokine; eosinophil chemotactic factor-L; Secreted protein Ym1
Gene Aliases: AI505981; Chi3l3; Chil3; ECF-L; Ym1
UniProt ID: (Mouse) O35744
Entrez Gene ID: (Mouse) 12655
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.