OriGene
Manufacturer Code:CF811213
Catalog # 50-167-0526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D8 |
Immunogen | Full length human recombit protein of human CHIA produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acidic mammalian chitinase; AMCase; AMCASE CHIT2 TSA1902; Lung-specific protein TSA1902
Gene Aliases: AMCASE; CHIA; CHIT2; TSA1902
UniProt ID: (Human) Q9BZP6
Entrez Gene ID: (Human) 27159
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.