Invitrogen
Manufacturer Code:PA576096
Catalog # PIPA576096
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human CHIA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acidic chitinase; Acidic mammalian chitinase; Acidic mammalian chitinase AMCase Lung-specific protein TSA1902 CHIA; AMCase; eosinophil chemotactic cytokine; Lung-specific protein TSA1902; small acid mammalian chitinase; YNL
Gene Aliases: 2200003E03Rik; AMCASE; CHIA; Chia1; CHIT2; TSA1902; YNL
UniProt ID: (Human) Q9BZP6, (Rat) Q6RY07, (Mouse) Q91XA9
Entrez Gene ID: (Human) 27159, (Rat) 113901, (Mouse) 81600
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.