Proteintech
Manufacturer Code:214841AP150UL
Catalog # 50-173-3444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHIA Fusion Protein Ag15767 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2200003E03Rik Acidic mammalian chitinase AMCase CHIA CHIT2 chitinase acidic Lung specific protein TSA1902 RP5 1125M8.1 TSA1902; acidic chitinase; Acidic mammalian chitinase; AMCase; eosinophil chemotactic cytokine; Lung-specific protein TSA1902; small acid mammalian chitinase; YNL
Gene Aliases: 2200003E03Rik; AMCASE; CHIA; Chia1; CHIT2; TSA1902; YNL
UniProt ID: (Human) Q9BZP6, (Rat) Q6RY07, (Mouse) Q91XA9
Entrez Gene ID: (Human) 27159, (Rat) 113901, (Mouse) 81600
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.