Invitrogen
Manufacturer Code:PA552144
Catalog # PIPA552144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CHI3L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chitinase 3-like 2; Chitinase-3-like protein 2; Chondrocyte protein 39; YKL-39; YKL-39 YKL39 chitinase-3-like protein 2 chondrocyte protein 39
Gene Aliases: CHI3L2; CHIL2; YKL-39; YKL39
UniProt ID: (Human) Q15782
Entrez Gene ID: (Human) 1117
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.