Invitrogen
Manufacturer Code:PA5112920
Catalog # PIPA5112920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Chitinase-3-like protein 2 protein (27-390AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHI3L2 CHIL2 chitinase 3 like 2 chitinase 3-like 2 Chitinase-3-like protein 2 Chondrocyte protein 39 YKL39 YKL-39; chitinase 3-like 2; Chitinase-3-like protein 2; Chondrocyte protein 39; YKL-39
Gene Aliases: CHI3L2; CHIL2; YKL-39; YKL39
UniProt ID: (Human) Q15782
Entrez Gene ID: (Human) 1117
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.