Invitrogen
Manufacturer Code:PA579042
Catalog # PIPA579042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse CHI3L1 recombit protein (Position: Y30-K262). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cartilage glycoprotein 39; CGP-39; chitinase 3-like 1 protein; Chitinase-3-like protein 1; Chitinase-3-like protein 1 BRP39 protein Breast regression protein 39 Cartilage glycoprotein 39 CGP-39 GP-39 Chi3l1 Brp39 Chil1; GP-39
Gene Aliases: Chi3l1; Chil1
UniProt ID: (Rat) Q9WTV1
Entrez Gene ID: (Rat) 89824
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.