Invitrogen
Manufacturer Code:PA547363
Catalog # PIPA547363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Chitinase 3-like 1 Tyr22-Ala381 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Breast regression protein 39; BRP39 protein; Cartilage glycoprotein 39; CGP-39; chitinase 3-like 1; Chitinase-3-like protein 1; Chitinase-3-like protein 1 39 kDa synovial protein Cartilage glycoprotein 39 CGP-39 GP-39 hCGP-39 YKL-40 CHI3L1 CGP39 GP39 hCGP39; GP-39
Gene Aliases: AW208766; Brp39; Chi3l1; Chil1; Gp39
UniProt ID: (Mouse) Q61362
Entrez Gene ID: (Mouse) 12654
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.