Invitrogen
Manufacturer Code:PA543746
Catalog # PIPA543746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CHI3L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa heparin-binding glycoprotein; 39 kDa synovial protein; 39 kDa whey protein; 40 kDa secretory signalling glycoprotein; BP40; Cartilage glycoprotein 39; cartilage glycoprotein-39; CGP-39; chitinase 3-like 1 (cartilage glycoprotein-39); Chitinase 3-like 1 (cartilage glycoprotein-39) DKFZp686N19119 FLJ38139 GP39 HC-gp39 HCGP-3P YKL40 YYL-40 ASRT7 GP-39 CGP-39 YKL-40 hCGP-39; chitinase 3-like 1 protein; Chitinase-3-like protein 1; Chitinase-like protein 1; CLP-1; GP-39; gp38k; Mammary gland protein 40; MGP-40; signal processing protein; Signal-processing protein; SPC-40; YKL-40
Gene Aliases: ASRT7; BP40; CGP-39; CHI3L1; CHIL1; CLP-1; GP-39; GP38K; GP39; HC-gp39; hCGP-39; HCGP-3P; MGP-40; SPC-40; SPS-40; YKL-40; YKL40; YYL-40
UniProt ID: (Goat) Q8SPQ0, (Bovine) P30922, (Human) P36222, (Pig) Q29411, (Rat) Q9WTV1
Entrez Gene ID: (Rabbit) 100338424, (Goat) 100860825, (Dog) 490222, (Bovine) 286869, (Human) 1116, (Pig) 396865, (Rat) 89824
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.