Invitrogen
Manufacturer Code:PA5115013
Catalog # PIPA5115013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CHI3L1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 39 kDa synovial protein; 39 kDa synovial protein ASRT7 AW208766 Breast regression protein 39 Brp39 BRP39 protein Cartilage glycoprotein 39 CGP39 CGP-39 Chi3l1 Chil1 chitinase 3 like 1 Chitinase 3 like protein 1CGP 39 chitinase 3-like 1 chitinase 3-like 1 (cartilage glycoprotein-39) chitinase 3-like 1 protein chitinase-3-like protein 1 chitinase-like 1 DKFZp686N19119 FLJ38139 Gp39 GP-39 hCGP39 HC-gp39 hCGP-39 HCGP-3P YKL40 YKL-40 YYL-40; Breast regression protein 39; BRP39 protein; Cartilage glycoprotein 39; CGP-39; chitinase 3-like 1; chitinase 3-like 1 (cartilage glycoprotein-39); chitinase 3-like 1 protein; Chitinase-3-like protein 1; GP-39; YKL-40
Gene Aliases: ASRT7; AW208766; Brp39; CGP-39; CHI3L1; Chil1; GP-39; GP39; HC-gp39; hCGP-39; HCGP-3P; YKL-40; YKL40; YYL-40
UniProt ID: (Human) P36222, (Rat) Q9WTV1, (Mouse) Q61362
Entrez Gene ID: (Human) 1116, (Rat) 89824, (Mouse) 12654
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.