OriGene
Manufacturer Code:TA506121
Catalog # 50-167-6470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B7 |
Immunogen | Full length human recombit protein of human CHGA produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AL-11; betagranin (N-terminal fragment of chromogranin A); Catestatin; CGA; chromofungin; Chromogranin-A; EA-92; ER-37; ES-43; GE-25; GR-44; GV-19; LF-19; p-Glu serpinin precursor; Pancreastatin; parathyroid secretory protein 1; Pituitary secretory protein I; Serpinin; Serpinin-RRG; SL21; SP-I; SS-18; Vasostatin I; Vasostatin II; Vasostatin-1; Vasostatin-2; WA-8; WE-14
Gene Aliases: CGA; CHGA
UniProt ID: (Human) P10645
Entrez Gene ID: (Human) 1113
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.