Invitrogen
Manufacturer Code:PA530655
Catalog # PIPA530655
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 351 and 538 of Human CHFR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Checkpoint with forkhead and RING finger domains protein; E3 ubiquitin-protein ligase CHFR; RING finger protein 196; RING-type E3 ubiquitin transferase CHFR; RNF116 RNF196 E3 ubiquitin-protein ligase CHFR RING finger protein 196 checkpoint with forkhead and RING finger domains protein
Gene Aliases: CHFR; RNF116; RNF196
UniProt ID: (Human) Q96EP1
Entrez Gene ID: (Human) 55743
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.