Invitrogen
Manufacturer Code:PA5106807
Catalog # PIPA5106807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CHFR corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5730484M20Rik C230082M18 checkpoint with forkhead and ring finger domains Checkpoint with forkhead and RING finger domains protein checkpoint with forkhead and ring finger domains E3 ubiquitin protein ligase Chfr E3 ubiquitin-protein ligase CHFR FLJ10796 FLJ33629 RING finger protein 196 RING-type E3 ubiquitin transferase CHFR RNF116 RNF196; Checkpoint with forkhead and RING finger domains protein; E3 ubiquitin-protein ligase CHFR; RING finger protein 196; RING-type E3 ubiquitin transferase CHFR
Gene Aliases: 5730484M20Rik; C230082M18; CHFR; RNF116; RNF196
UniProt ID: (Human) Q96EP1, (Mouse) Q810L3
Entrez Gene ID: (Human) 55743, (Rat) 288734, (Mouse) 231600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.