Novus Biologicals
Manufacturer Code:NBP130986
Catalog # NBP130986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CHFR. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: checkpoint with forkhead and ring finger domains Checkpoint with forkhead and RING finger domains protein E3 ubiquitin-protein ligase CHFR EC 6.3.2.- FLJ10796 FLJ33629 RING finger protein 196 RNF196RNF116; Checkpoint with forkhead and RING finger domains protein; E3 ubiquitin-protein ligase CHFR; RING finger protein 196; RING-type E3 ubiquitin transferase CHFR
Gene Aliases: CHFR; RNF116; RNF196
UniProt ID: (Human) Q96EP1
Entrez Gene ID: (Human) 55743
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.