Abnova
Manufacturer Code:PAB5293
Catalog # 89-131-799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding T68 of human CHEK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CHEK2.
For Research Use Only
Protein Aliases: CDS1; Cds1 homolog; Checkpoint kinase 2; checkpoint-like protein CHK2; CHK2; CHK2 checkpoint homolog; HuCds1; LFS2; PP1425; RAD53; Serine/threonine-protein kinase Chk2
Gene Aliases: CDS1; CHEK2; CHK2; hCds1; HuCds1; LFS2; PP1425; RAD53
UniProt ID: (Human) O96017
Entrez Gene ID: (Human) 11200
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.