Invitrogen
Manufacturer Code:PA5100229
Catalog # PIPA5100229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CHDH corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU041472 CDH CHD CHDH choline dehydrogenase choline dehydrogenase mitochondrial D630034H06Rik; CDH; CHD; Choline dehydrogenase, mitochondrial
Gene Aliases: AU041472; CDH; CHD; CHDH; D630034H06Rik
UniProt ID: (Human) Q8NE62, (Rat) Q6UPE0, (Mouse) Q8BJ64
Entrez Gene ID: (Human) 55349, (Rat) 290551, (Mouse) 218865
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.