Invitrogen
Manufacturer Code:PA572690
Catalog # PIPA572690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHD7 antibody for an 18 amino acid synthetic peptide near the amino terminus of human CHD7. The immunogen is within amino acids 670 - 720 of CHD7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase CHD7; CHARGE association; CHD-7; chromodomain helicase DNA binding protein 7 isoform CRA_e; Chromodomain-helicase-DNA-binding protein 7; HH5 IS3 KAL5 ATP-dependent helicase CHD7 chromodomain helicase DNA binding protein 7 isoform CRA&lowbare chromodomain-helicase-DNA-binding protein 7
Gene Aliases: A730019I05Rik; CHD7; CRG; Cycn; Cyn; Dz; Edy; Flo; GENA 47; Gena 52; GENA 60; HH5; IS3; KAL5; KIAA1416; Lda; metis; Mt; Obt; Todo; WBE1; Whi
UniProt ID: (Human) Q9P2D1, (Mouse) A2AJK6
Entrez Gene ID: (Human) 55636, (Mouse) 320790
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.