Invitrogen
Manufacturer Code:PA581154
Catalog # PIPA581154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the Human CHD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase CHD4; CHD-4; Chromodomain-helicase-DNA-binding protein 4; Mi-2 autoantigen 218 kDa protein; Mi-2b Mi2-BETA ATP-dependent helicase CHD4 CHD-4 Mi-2 autoantigen 218 kDa protein chromodomain-helicase-DNA-binding protein 4 MI2B; Mi2-beta
Gene Aliases: CHD-4; CHD4; Mi-2b; Mi2-BETA
UniProt ID: (Human) Q14839
Entrez Gene ID: (Human) 1108
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.