Novus Biologicals
Manufacturer Code:NBP299099C
Catalog # NB084094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human CHD4. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase CHD4; ATP-dependent helicase CHD4 CHD-4 chromodomain helicase DNA binding protein 4 chromodomain-helicase-DNA-binding protein 4 DKFZp686E06161 EC 3.6.1 EC 3.6.4.12 Mi-2 autoantigen 218 kDa protein Mi-2b Mi2-BETA; CHD-4; Chromodomain-helicase-DNA-binding protein 4; Mi-2 autoantigen 218 kDa protein; Mi2-beta
Gene Aliases: CHD-4; CHD4; Mi-2b; Mi2-BETA
UniProt ID: (Human) Q14839
Entrez Gene ID: (Human) 1108
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.