Novus Biologicals
Manufacturer Code:NBP257060
Catalog # NBP257060
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MSYCRQEGKDRIIFVTKEDHETPSSAE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHDH4 coiled-coil-helix-coiled-coil-helix domain containing 4 Coiled-coil-helix-coiled-coil-helix domain-containing protein 4 FLJ31709 MIA40mitochondrial intermembrane space import and assembly protein 40 mitochondrial intermembrane space import and assembly 40 homolog TIMM40 translocase of inner mitochondrial membrane 40 homolog; Coiled-coil-helix-coiled-coil-helix domain-containing protein 4; mitochondrial intermembrane space import and assembly 40 homolog; Mitochondrial intermembrane space import and assembly protein 40; translocase of inner mitochondrial membrane 40 homolog
Gene Aliases: CHCHD4; MIA40; TIMM40
UniProt ID: (Human) Q8N4Q1
Entrez Gene ID: (Human) 131474
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.