Proteintech
Manufacturer Code:256711AP150UL
Catalog # 50-100-2979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHCHD10 Fusion Protein Ag22598 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C22orf16 CHCHD10 N27C7 4 Protein N27C7 4; Coiled-coil-helix-coiled-coil-helix domain-containing protein 10, mitochondrial; Nur77 downstream gene 2; Nur77 downstream protein 2; Protein N27C7-4
Gene Aliases: 1620401E04Rik; AI267078; C22orf16; CHCHD10; FTDALS2; IMMD; N27C7-4; Ndg2; SMAJ
UniProt ID: (Human) Q8WYQ3
Entrez Gene ID: (Human) 400916, (Mouse) 103172
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.