Abnova
Manufacturer Code:PAB14525
Catalog # 89-117-117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified CHAT from human placenta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against native CHAT.
For Research Use Only
Protein Aliases: acetyl CoA:choline O-acetyltransferase; CHOACTase; choline acetylase; Choline O-acetyltransferase; CMS1A; CMS1A2
Gene Aliases: CHAT; CHOACTASE; CMS1A; CMS1A2; CMS6
UniProt ID: (Human) P28329
Entrez Gene ID: (Human) 1103
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.