Proteintech
Manufacturer Code:170371AP150UL
Catalog # 50-173-1342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHAF1A Fusion Protein Ag10536 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAF CAF 1 CAF 1 subunit A CAF I 150 kDa subunit CAF I p150 CAF1 CAF1B CAF1P150 CHAF1A hp150 P150; CAF-1 subunit A; CAF-I 150 kDa subunit; CAF-I p150; CAF-Ip150; Chromatin assembly factor 1 subunit A; chromatin assembly factor 1, subunit A (p150); chromatin assembly factor I (150 kDa); Chromatin assembly factor I p150 subunit; CTB-50L17.7; hp150
Gene Aliases: AL023013; AL024058; Cac1p; CAF; CAF-1; CAF1; CAF1B; CAF1P150; Caip150; CHAF1A; P150
UniProt ID: (Human) Q13111, (Mouse) Q9QWF0
Entrez Gene ID: (Human) 10036, (Mouse) 27221
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.