Proteintech
Manufacturer Code:129631AP150UL
Catalog # 50-172-8382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHAD Fusion Protein Ag4077 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cartilage leucine rich protein CHAD chondroadherin SLRR4A; Cartilage leucine-rich protein; Chondroadherin; chondroadherin proteoglycan
Gene Aliases: CHAD; SLRR4A
UniProt ID: (Human) O15335, (Mouse) O55226, (Rat) O70210
Entrez Gene ID: (Human) 1101, (Mouse) 12643, (Rat) 29195
Molecular Function: cytokine extracellular matrix protein receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.