Invitrogen
Manufacturer Code:PA136017
Catalog # 01-672-207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic human CGRP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-type CGRP; Calca CA; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin-related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; CCP; CGRP-I; Katacalcin; PDN-21
Gene Aliases: Cal1; CAL6; Calc; CALC1; CALCA; calcitonin; CGRP; CGRP-I; CGRP1; CT; KC; PCT; RATCAL6
UniProt ID: (Human) P06881, (Rat) P01256
Entrez Gene ID: (Human) 796, (Rat) 24241
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.