Invitrogen
Manufacturer Code:PA542181
Catalog # PIPA542181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ABHD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; 1-acylglycerol-3-phosphate O-acyltransferase ABHD5-like; abhydrolase domain containing 5; Abhydrolase domain containing 5 CDS CGI58 IECN2 MGC8731 NCIE2; Abhydrolase domain-containing protein 5; abhydrolase domain-containing protein 5 variant 1; abhydrolase domain-containing protein 5 variant 2; alpha/beta hydrolase domain-containing 5; lipid droplet binding protein; Lipid droplet-binding protein CGI-58; Protein CGI-58; truncated abhydrolase domain-containing protein 5
Gene Aliases: 1300003D03Rik; 2010002J10Rik; ABHD5; CDS; CGI-58; CGI58; IECN2; IECN5; NCIE2
UniProt ID: (Human) Q8WTS1, (Pig) Q5EE05, (Mouse) Q9DBL9, (Rat) Q6QA69
Entrez Gene ID: (Dog) 485570, (Human) 51099, (Sheep) 100125355, (Rabbit) 100350162, (Horse) 100066775, (Pig) 497624, (Bovine) 535588, (Guinea pig) 100735683, (Mouse) 67469, (Rat) 316122
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.