Novus Biologicals
Manufacturer Code:NB300511SS
Catalog # NB300511SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CF3 |
Immunogen | Synthetic Peptide: G(103) R I I A S Y D P D N K E E R(117) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABC35 ABCC7CF ATP-binding cassette sub-family C member 7 ATP-binding cassette transporter sub-family C member 7 cAMP-dependent chloride channel CFTR/MRP Channel conductance-controlling ATPase cystic fibrosis transmembrane conductance regulator cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C member 7) cystic fibrosis transmembrane conductance regulator ATP-binding cassette(sub-family C member 7) dJ760C5.1 EC 3.6.3 MRP7EC 3.6.3.49 TNR-CFTR; ATP-binding cassette sub-family C member 7; cAMP-dependent chloride channel; CFTR; Channel conductance-controlling ATPase; Cystic fibrosis transmembrane conductance regulator; cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7)
Gene Aliases: ABC35; ABCC7; CF; CFTR; CFTR/MRP; dJ760C5.1; MRP7; TNR-CFTR
UniProt ID: (Human) P13569
Entrez Gene ID: (Human) 1080
Molecular Function:
ATP-binding cassette (ABC) transporter
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.