Proteintech
Manufacturer Code:207381AP150UL
Catalog # 50-173-3056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC35 ABCC7 CF CFTR CFTR/MRP MRP7 TNR CFTR; ATP-binding cassette sub-family C member 7; cAMP-dependent chloride channel; CFTR; Channel conductance-controlling ATPase; Cystic fibrosis transmembrane conductance regulator; cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7)
Gene Aliases: ABC35; ABCC7; CF; CFTR; CFTR/MRP; dJ760C5.1; MRP7; TNR-CFTR
UniProt ID: (Human) P13569
Entrez Gene ID: (Human) 1080
Molecular Function:
ATP-binding cassette (ABC) transporter
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.