Proteintech
Manufacturer Code:171921AP150UL
Catalog # 50-173-1435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CFP Fusion Protein Ag9798 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BFD CFP Complement factor P complement factor properdin PFC PFD PROPERDIN; Complement factor P; Properdin; properdin P factor, complement
Gene Aliases: BFD; CFP; PFC; PFD; PROPERDIN
UniProt ID: (Human) P27918
Entrez Gene ID: (Human) 5199
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.