OriGene
Manufacturer Code:CF804552
Catalog # 50-166-7030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B9 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-272 of human CFLAR produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-FLIP; CASH; CASH CASP8AP1 CLARP Casper FLAME FLAME-1 FLAME1 FLIP I-FLICE MRIT c-FLIP c-FLIPL c-FLIPR c-FLIPS; CASP8 and FADD-like apoptosis regulator; CASP8 and FADD-like apoptosis regulator subunit p12; CASP8 and FADD-like apoptosis regulator subunit p43; Caspase homolog; Caspase-eight-related protein; Caspase-like apoptosis regulatory protein; caspase-related inducer of apoptosis; Casper; Cellular FLICE-like inhibitory protein; CLARP; FADD-like antiapoptotic molecule 1; FLAME-1; I-FLICE; Inhibitor of FLICE; MACH-related inducer of toxicity; MRIT; testis secretory sperm-binding protein Li 233m; Usurpin; usurpin beta
Gene Aliases: c-FLIP; c-FLIPL; c-FLIPR; c-FLIPS; CASH; CASP8AP1; Casper; CFLAR; CLARP; FLAME; FLAME-1; FLAME1; FLIP; I-FLICE; MRIT
UniProt ID: (Human) O15519
Entrez Gene ID: (Human) 8837
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.