Invitrogen
Manufacturer Code:MA531677
Catalog # PIMA531677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E1E10 |
Immunogen | Purified recombit fragment of human CFHR5 (AA: 344-569) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: complement factor H-related 5; Complement factor H-related protein 5; factor H-related protein 5; FHR-5; FHR5 CFHL5 FHR-5 CFHR5D
Gene Aliases: CFHL5; CFHR5; CFHR5D; FHR-5; FHR5
UniProt ID: (Human) Q9BXR6
Entrez Gene ID: (Human) 81494
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.