Abnova
Manufacturer Code:PAB20088
Catalog # 89-121-589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human CFB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant CFB.
For Research Use Only
Protein Aliases: B-factor, properdin; BF; BFD; C3 proaccelerator; C3 proactivator; C3/C5 convertase; CFAB; Complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; FB; FBI12; GBG; Glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; H2-Bf; PBF2; Properdin factor B
Gene Aliases: AHUS4; ARMD14; BF; BFD; CFAB; CFB; CFBD; FB; FBI12; GBG; H2-Bf; PBF2
UniProt ID: (Human) P00751
Entrez Gene ID: (Human) 629
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.